Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079711.1 | NP_001073179.1 | 5 | 563 | ||
XM_006237502.2 | XP_006237564.1 | 5 | 543 | |||
XM_006237504.2 | XP_006237566.1 | 5 | 557 | |||
GenBank mRNA | BC128747.1 | - | 5 | 563 | 7099220 |