Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324309.1 | NP_001311238.1 | 2 | 1387 | ||
NM_014851.3 | NP_055666.2 | 2 | 1387 | |||
GenBank mRNA | AB007938.1 | - | 2 | 1490 | ||
AK075305.1 | - | 2 | 1358 | |||
AK090472.1 | - | 2 | 1156 | |||
AK307564.1 | - | 2 | 347 | |||
BC034039.1 | - | 2 | 1358 | 4811534 | ||
BC064980.1 | - | 2 | 1340 | |||
BC091648.1 | - | 2 | 1340 | 6373903 |