Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105712.1 | NP_001099182.1 | 2 | 136 | ||
XM_008762681.1 | XP_008760903.1 | 1 | 1829 | |||
XM_008762682.1 | XP_008760904.1 | 2 | 612 | |||
XM_008762683.1 | XP_008760905.1 | 1 | 1829 | |||
GenBank mRNA | M92076.1 | - | 1 | 54 |
Set Membership: |
G-protein coupled receptor |