Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014855.2 | NP_055670.1 | 7 | 1001 | ||
XM_017012864.1 | XP_016868353.1 | 6 | 828 | |||
GenBank mRNA | AB007875.1 | - | 7 | 980 | ||
BC037399.1 | - | 5 | 674 | 5172434 | ||
BQ228738.1 | - | 27 | ||||
CD671081.1 | - | 4 | 425 |