Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014855.2 | NP_055670.1 | 16 | 2119 | ||
XM_017012864.1 | XP_016868353.1 | 15 | 1946 | |||
GenBank mRNA | AB007875.1 | - | 16 | 2343 | ||
AK092068.1 | - | 3 | 1582 | |||
BC008841.2 | - | 7 | 787 | |||
BC014240.2 | - | 3 | 310 | |||
BC037399.1 | - | 13 | 2321 | 5172434 |