Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012297.4 | NP_036429.2 | 12 | 1504 | ||
NM_203504.2 | NP_987100.1 | 11 | 1540 | |||
NM_203505.2 | NP_987101.1 | 12 | 1639 | |||
XM_005263382.2 | XP_005263439.1 | 12 | 1613 | |||
XM_005263383.3 | XP_005263440.1 | 13 | 1578 | |||
XM_011532441.2 | XP_011530743.1 | 12 | 1499 | |||
XM_017008876.1 | XP_016864365.1 | 12 | 1487 | |||
XM_017008877.1 | XP_016864366.1 | 12 | 1666 | |||
XM_017008878.1 | XP_016864367.1 | 11 | 1388 | |||
XM_017008879.1 | XP_016864368.1 | 11 | 1414 | |||
GenBank mRNA | AB014560.1 | - | 12 | 1459 | ||
AF051311.1 | - | 10 | 1240 | |||
AF053535.1 | - | 11 | 1435 | |||
AF070615.1 | - | 8 | 939 | |||
AF145284.1 | - | 12 | 1534 | |||
AK291772.1 | - | 11 | 1470 | |||
AK291786.1 | - | 12 | 1474 | |||
BC011731.2 | - | 11 | 1458 | 4337457 |