Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014856.2 | NP_055671.2 | 10 | 1881 | ||
XM_005245678.4 | XP_005245735.1 | 10 | 2038 | |||
XM_005245679.2 | XP_005245736.1 | 10 | 1888 | |||
XM_011510219.1 | XP_011508521.1 | 9 | 1503 | |||
XM_011510220.2 | XP_011508522.1 | 9 | 1701 | |||
XM_011510221.2 | XP_011508523.1 | 8 | 1409 | |||
XM_011510222.1 | XP_011508524.1 | 9 | 1503 | |||
XM_017002993.1 | XP_016858482.1 | 7 | 1263 | |||
GenBank mRNA | AB007945.2 | - | 10 | 1881 | ||
AK226053.1 | - | 5 | 530 |