Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014856.2 | NP_055671.2 | 13 | 2288 | ||
XM_005245678.4 | XP_005245735.1 | 13 | 2445 | |||
XM_005245679.2 | XP_005245736.1 | 13 | 2295 | |||
XM_011510219.1 | XP_011508521.1 | 11 | 1694 | |||
XM_011510220.2 | XP_011508522.1 | 12 | 2108 | |||
XM_011510221.2 | XP_011508523.1 | 11 | 1816 | |||
XM_011510222.1 | XP_011508524.1 | 12 | 1910 | |||
XM_017002993.1 | XP_016858482.1 | 10 | 1670 | |||
GenBank mRNA | AB007945.2 | - | 13 | 2288 | ||
AK226053.1 | - | 8 | 937 |