Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282729.1 | NP_001269658.1 | 4 | 1183 | ||
NM_001282730.1 | NP_001269659.1 | 4 | 1391 | |||
NM_001282731.1 | NP_001269660.1 | 4 | 1233 | |||
NM_001282732.1 | NP_001269661.1 | 3 | 773 | |||
NM_014860.2 | NP_055675.1 | 4 | 1178 | |||
XM_005264672.4 | XP_005264729.1 | 4 | 1380 | |||
GenBank mRNA | AB018307.1 | - | 4 | 920 | ||
AF197954.1 | - | 4 | 939 | |||
AF224759.1 | - | 3 | 1768 | |||
AK000329.1 | - | 4 | 948 | |||
AK000616.1 | - | 4 | 734 | |||
AK304889.1 | - | 3 | 556 | |||
BC074000.1 | - | 4 | 920 | 6501232 | ||
CR456983.1 | - | 2 | 577 |