Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282729.1 | NP_001269658.1 | 3 | 962 | ||
NM_001282730.1 | NP_001269659.1 | 3 | 1170 | |||
NM_001282731.1 | NP_001269660.1 | 3 | 1012 | |||
NM_014860.2 | NP_055675.1 | 3 | 957 | |||
XM_005264672.4 | XP_005264729.1 | 3 | 1159 | |||
GenBank mRNA | AB018307.1 | - | 3 | 699 | ||
AF197954.1 | - | 3 | 718 | |||
AF224759.1 | - | 2 | 1547 | |||
AK000329.1 | - | 3 | 727 | |||
AK000616.1 | - | 3 | 513 | |||
BC074000.1 | - | 3 | 699 | 6501232 | ||
BI755200.1 | - | 3 | 514 | |||
CR456983.1 | - | 1 | 356 |