Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014865.3 | NP_055680.3 | 9 | 1007 | ||
GenBank mRNA | AK125155.1 | - | 6 | 662 | ||
AK128354.1 | - | 9 | 919 | |||
AK303310.1 | - | 8 | 982 | |||
BC016913.1 | - | 9 | 1739 | |||
BC028182.1 | - | 9 | 1019 | 5395775 | ||
D63880.1 | - | 9 | 1760 |