Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC689757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101019.2 | NP_001094489.2 | 4 | 641 | ||
XM_008763332.1 | XP_008761554.1 | 4 | 944 | |||
GenBank mRNA | EF100687.1 | - | 4 | 611 | ||
FJ404472.1 | - | 4 | 641 |