Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085404.1 | NP_001078873.1 | 4 | 563 | ||
XM_006237453.2 | XP_006237515.1 | 5 | 596 | |||
XM_008763333.1 | XP_008761555.1 | 4 | 1622 | |||
GenBank mRNA | EF100689.1 | - | 4 | 563 |