Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127460.2 | NP_001120932.1 | 25 | 3827 | ||
NM_001166279.1 | NP_001159751.1 | 25 | 3753 | |||
NM_014871.4 | NP_055686.3 | 25 | 3744 | |||
GenBank mRNA | AB014610.2 | - | 25 | 3650 | ||
AB107585.1 | - | 24 | 3438 | |||
AK001232.1 | - | 9 | 1012 | |||
AK096679.1 | - | 12 | 1815 | |||
BC024043.1 | - | 25 | 3657 | 3357890 | ||
BC094885.1 | - | 25 | 3824 | 4823602 | ||
BX648106.1 | - | 24 | 3855 |