Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109552.2 | NP_001103022.1 | 3 | 657 | ||
XM_008768114.1 | XP_008766336.1 | 2 | 752 | |||
GenBank mRNA | FM055064.1 | - | 3 | 610 |