Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022514.1 | NP_071959.1 | 2 | 238 | ||
GenBank mRNA | BC058474.1 | - | 3 | 268 | 6919338 | |
BC091566.1 | - | 3 | 236 | 7388530 | ||
FQ210011.1 | - | 3 | 254 | |||
FQ214023.1 | - | 3 | 258 | |||
FQ217713.1 | - | 2 | 178 | |||
FQ221292.1 | - | 3 | 263 | |||
FQ221643.1 | - | 3 | 255 | |||
FQ221684.1 | - | 3 | 256 | |||
FQ221938.1 | - | 3 | 269 | |||
FQ222018.1 | - | 3 | 258 | |||
FQ222606.1 | - | 3 | 259 | |||
FQ222991.1 | - | 3 | 260 | |||
FQ223942.1 | - | 2 | 187 | |||
FQ224664.1 | - | 3 | 256 | |||
FQ228505.1 | - | 3 | 255 | |||
FQ228603.1 | - | 3 | 256 | |||
FQ228786.1 | - | 3 | 255 | |||
X07424.1 | - | 2 | 238 |