Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305792.1 | NP_001292721.1 | 7 | 1864 | ||
NM_014875.2 | NP_055690.1 | 9 | 2276 | |||
XM_011510231.2 | XP_011508533.1 | 10 | 2370 | |||
XM_011510232.2 | XP_011508534.1 | 9 | 2032 | |||
XM_011510233.2 | XP_011508535.1 | 10 | 2163 | |||
XM_011510235.2 | XP_011508537.1 | 10 | 1875 | |||
XM_011510236.1 | XP_011508538.1 | 8 | 2159 | |||
XM_017003005.1 | XP_016858494.1 | 8 | 2453 | |||
XM_017003006.1 | XP_016858495.1 | 9 | 2061 | |||
XM_017003007.1 | XP_016858496.1 | 9 | 1588 | |||
GenBank mRNA | BC113742.1 | - | 8 | 1915 | 8322794 | |
D26361.1 | - | 9 | 2276 |