Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079531.1 | NP_001072999.1 | 4 | 398 | ||
GenBank mRNA | AY325236.1 | - | 4 | 398 |