Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198763.1 | NP_001185692.1 | 3 | 324 | ||
NM_001198764.1 | NP_001185693.1 | 2 | 213 | |||
NM_014880.4 | NP_055695.2 | 3 | 324 | |||
GenBank mRNA | AF499000.1 | - | 3 | 278 | ||
AK291279.1 | - | 3 | 277 | |||
AK304422.1 | - | 3 | 311 | |||
AY314007.1 | - | 3 | 311 | |||
BC020646.1 | - | 2 | 160 | 4716449 | ||
D14664.1 | - | 3 | 265 | |||
DB217404.1 | - | 3 | 324 | |||
DB460994.1 | - | 2 | 180 |
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 36 | 5264 | ||
NM_001198760.1 | NP_001185689.1 | 35 | 5096 | |||
GenBank mRNA | AY184222.1 | - | 36 | 5190 | ||
AY314006.1 | - | 35 | 5022 |