Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007231.2 | NP_001007232.2 | 2 | 649 | ||
NM_001166276.1 | NP_001159748.1 | 1 | 589 | |||
NM_001166277.1 | NP_001159749.1 | 2 | 649 | |||
NM_014882.2 | NP_055697.1 | 1 | 589 | |||
XM_005264675.2 | XP_005264732.1 | 2 | 356 | |||
XM_005264676.2 | XP_005264733.1 | 2 | 354 | |||
XM_011533206.2 | XP_011531508.1 | 2 | 397 | |||
XM_011533207.2 | XP_011531509.1 | 2 | 363 | |||
XM_011533208.2 | XP_011531510.1 | 3 | 414 | |||
XM_017005423.1 | XP_016860912.1 | 2 | 257 | |||
XM_017005424.1 | XP_016860913.1 | 2 | 257 | |||
XM_017005425.1 | XP_016860914.1 | 2 | 257 | |||
XM_017005428.1 | XP_016860917.1 | 2 | 257 | |||
GenBank mRNA | AK290396.1 | - | 2 | 327 | ||
AK297056.1 | - | 2 | 527 | |||
AK297157.1 | - | 1, 2 | 392 | |||
AK298669.1 | - | 1 | 416 | |||
AK303591.1 | - | 3 | 484 | |||
AK310742.1 | - | 2 | 354 | |||
BC039591.1 | - | 1 | 450 | 5757311 | ||
D29642.1 | - | 1 | 402 | |||
EU794656.1 | - | 1 | 589 |