Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005110.3 | NP_005101.1 | 15 | 1651 | ||
XM_006714942.2 | XP_006715005.1 | 15 | 1600 | |||
XM_017010100.1 | XP_016865589.1 | 14 | 1536 | |||
GenBank mRNA | AB016789.1 | - | 15 | 1580 | ||
AK001242.1 | - | 5 | 533 | |||
AK022507.1 | - | 15 | 1591 | |||
AK297750.1 | - | 13 | 1287 | |||
AK314551.1 | - | 15 | 1591 | |||
BC000012.2 | - | 15 | 1589 | 3506723 | ||
BT009818.1 | - | 14 | 1481 | |||
KU178494.1 | - | 14 | 1481 | |||
KU178495.1 | - | 14 | 1481 |