Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005110.3 | NP_005101.1 | 3 | 304 | ||
XM_006714942.2 | XP_006715005.1 | 3 | 253 | |||
XM_017010100.1 | XP_016865589.1 | 3 | 251 | |||
GenBank mRNA | AB016789.1 | - | 3 | 233 | ||
AK022507.1 | - | 3 | 244 | |||
AK314551.1 | - | 3 | 244 | |||
BC000012.2 | - | 3 | 242 | 3506723 | ||
BI753491.1 | - | 3 | 304 | |||
BT009818.1 | - | 2 | 134 | |||
KU178494.1 | - | 2 | 134 | |||
KU178495.1 | - | 2 | 134 |