Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC690483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228950.2 | XP_006229012.1 | 5 | 1632 | ||
GenBank mRNA | BC082038.1 | - | 4 | 1174 |