Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145858.2 | NP_665857.2 | 6 | 480 | ||
GenBank mRNA | AF029689.1 | - | 6 | 347 | ||
AF098798.1 | - | 6 | 347 | |||
AK001293.1 | - | 6 | 373 | |||
AK057604.1 | - | 6 | 964 | |||
AK315707.1 | - | 6 | 340 | |||
BC033023.2 | - | 6 | 334 | 5269248 | ||
BI599245.1 | - | 6 | 385 | |||
BX647276.1 | - | 5 | 322 | |||
BX648547.1 | - | 7 | 494 | |||
DB037192.1 | - | 6 | 481 |