Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145858.2 | NP_665857.2 | 8 | 714 | ||
GenBank mRNA | AF029689.1 | - | 8 | 581 | ||
AF098798.1 | - | 8 | 581 | |||
AK001293.1 | - | 8 | 607 | |||
AK057604.1 | - | 8 | 1198 | |||
AK058071.1 | - | 3 | 226 | |||
AK315707.1 | - | 8 | 574 | |||
BC033023.2 | - | 8 | 568 | 5269248 | ||
BX647276.1 | - | 7 | 556 | |||
BX648547.1 | - | 9 | 728 |