Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758871.1 | XP_008757093.1 | 1 | 696 |
Gene Symbol: LOC100910106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758830.1 | XP_008757052.1 | 3 | 796 |
Gene Symbol: Zfp758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228088.2 | XP_006228150.1 | 4 | 3118 |