Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005112.4 | NP_005103.2 | 11 | 1537 | ||
NM_017491.3 | NP_059830.1 | 14 | 1957 | |||
XM_017008880.1 | XP_016864369.1 | 14 | 2150 | |||
GenBank mRNA | AB010427.2 | - | 14 | 1876 | ||
AB209746.1 | - | 14 | 1730 | |||
AF020056.1 | - | 14 | 1785 | |||
AK127665.1 | - | 14 | 1697 | |||
AK222764.1 | - | 14 | 1829 | |||
AK222897.1 | - | 14 | 1861 | |||
AK225276.1 | - | 14 | 1829 | |||
AK291614.1 | - | 14 | 1807 | |||
AK302209.1 | - | 10 | 1229 | |||
AL050108.1 | - | 12 | 1505 | |||
AL110208.1 | - | 5 | 559 | |||
BC000201.1 | - | 14 | 1756 | 3351638 | ||
BC002489.2 | - | 14 | 1748 | 3051319 | ||
BC030541.2 | - | 11 | 1333 | 4403484 | ||
BX648190.1 | - | 10 | 2607 | |||
EU794675.1 | - | 14 | 1748 |