Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005112.4 | NP_005103.2 | 2 | 404 | ||
NM_017491.3 | NP_059830.1 | 2 | 404 | |||
XM_017008880.1 | XP_016864369.1 | 2 | 438 | |||
GenBank mRNA | AB010427.2 | - | 2 | 323 | ||
AB209746.1 | - | 2 | 177 | |||
AF020056.1 | - | 2 | 232 | |||
AF020260.1 | - | 2 | 232 | |||
AK127665.1 | - | 2 | 249 | |||
AK222764.1 | - | 2 | 276 | |||
AK222897.1 | - | 2 | 308 | |||
AK225276.1 | - | 2 | 276 | |||
AK291614.1 | - | 2 | 254 | |||
AK299602.1 | - | 2 | 249 | |||
AK303132.1 | - | 250 | ||||
BC000201.1 | - | 2 | 203 | 3351638 | ||
BC002489.2 | - | 2 | 195 | 3051319 | ||
BC030541.2 | - | 2 | 200 | 4403484 | ||
BI831941.1 | - | 2 | 405 | |||
EU794675.1 | - | 2 | 195 |