Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168664.1 | NP_001162135.1 | 50 | 6704 | ||
GenBank mRNA | BC169100.1 | - | 17 | 2570 |