Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005112.4 | NP_005103.2 | 7, 8 | 1059 | ||
NM_017491.3 | NP_059830.1 | 10, 11 | 1479 | |||
XM_017008880.1 | XP_016864369.1 | 10, 11 | 1672 | |||
GenBank mRNA | AB010427.2 | - | 10, 11 | 1398 | ||
AB209746.1 | - | 10, 11 | 1252 | |||
AF020056.1 | - | 10, 11 | 1307 | |||
AF020260.1 | - | 10, 11 | 1307 | |||
AK127665.1 | - | 10, 11 | 1219 | |||
AK222764.1 | - | 10, 11 | 1351 | |||
AK222897.1 | - | 10, 11 | 1383 | |||
AK225276.1 | - | 10, 11 | 1351 | |||
AK291614.1 | - | 10, 11 | 1329 | |||
AK302209.1 | - | 6, 7 | 751 | |||
AK303132.1 | - | 880 | ||||
AL050108.1 | - | 8, 9 | 1027 | |||
AL110208.1 | - | 1, 2 | 81 | |||
BC000201.1 | - | 10, 11 | 1278 | 3351638 | ||
BC002489.2 | - | 10, 11 | 1270 | 3051319 | ||
BC030541.2 | - | 7, 8 | 855 | 4403484 | ||
BX648190.1 | - | 6, 7 | 2129 | |||
EU794675.1 | - | 10, 11 | 1270 |