Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044301.1 | NP_001037766.1 | 2 | 429 | ||
XM_006234695.2 | XP_006234757.1 | 3 | 665 | |||
XM_008762107.1 | XP_008760329.1 | 2 | 442 | |||
XM_008762108.1 | XP_008760330.1 | 2 | 478 | |||
GenBank mRNA | BC083925.1 | - | 2 | 429 | 7135360 |