Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109621.1 | NP_001103091.1 | 2 | 827 | ||
GenBank mRNA | FQ212876.1 | - | 2 | 357 |