Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126098.1 | NP_001119570.1 | 4 | 748 | ||
XM_008765987.1 | XP_008764209.1 | 4 | 537 | |||
XM_008765988.1 | XP_008764210.1 | 4 | 574 | |||
XM_008765989.1 | XP_008764211.1 | 4 | 570 |