Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005121.2 | NP_005112.2 | 23 | 5346 | ||
XM_011525551.2 | XP_011523853.1 | 23 | 6047 | |||
XM_011525552.2 | XP_011523854.1 | 23 | 6047 | |||
XM_011525553.2 | XP_011523855.1 | 20 | 4761 | |||
GenBank mRNA | AB011165.2 | - | 20 | 4764 | ||
AF117754.1 | - | 24 | 5346 | |||
BC140891.1 | - | 23 | 5320 | 9021759 |