Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134846.1 | NP_001128318.1 | 1 | 546 |
Gene Symbol: LOC103690165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771605.1 | XP_008769827.1 | 1 | 3042 |