Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109640.1 | NP_001103110.1 | 5 | 656 | ||
XM_008761423.1 | XP_008759645.1 | 5 | 677 |