Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005127.2 | NP_005118.2 | 3, 4 | 902 | ||
GenBank mRNA | AB015628.1 | - | 3, 4 | 395 | ||
AK313916.1 | - | 3, 4 | 406 | |||
AY142147.1 | - | 3, 4 | 379 | |||
X96719.1 | - | 3, 4 | 377 |