Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320714.1 | NP_001307643.1 | 30, 31 | 6168 | ||
NM_005128.3 | NP_005119.2 | 30, 31 | 6059 | |||
XM_017028509.1 | XP_016883998.1 | 27, 28 | 5562 | |||
GenBank mRNA | AB023150.2 | - | 27, 28 | 5528 | ||
AJ237839.2 | - | 30, 31 | 6065 | |||
AK026478.1 | - | 9, 10 | 995 | |||
BC012791.2 | - | 12, 13 | 1680 |