Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032279.1 | NP_001027450.1 | 8 | 1432 | ||
NM_005133.2 | NP_005124.1 | 8 | 1436 | |||
GenBank mRNA | AL713740.1 | - | 5 | 2690 | ||
BC052622.1 | - | 8 | 1393 | |||
Y13835.2 | - | 8 | 1405 |