Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207000.1 | NP_001193929.1 | 3 | 1188 | ||
NM_031372.3 | NP_112740.1 | 3 | 1188 | |||
NR_003249.2 | 3 | 1188 | ||||
GenBank mRNA | AB017019.1 | - | 3 | 1188 | ||
AB066484.1 | - | 3 | 296 | |||
AY453824.1 | - | 3 | 287 | |||
BC007392.2 | - | 3 | 640 | |||
BC011714.2 | - | 3 | 829 | 3929324 | ||
BC018961.2 | - | 3 | 829 | |||
BC071944.1 | - | 3 | 769 | 5015862 | ||
CR407623.1 | - | 3 | 653 | |||
D89092.1 | - | 3 | 404 | |||
D89678.1 | - | 3 | 263 |