Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109650.1 | NP_001103120.1 | 1 | 675 | ||
XM_006252330.2 | XP_006252392.1 | 2 | 3225 |