Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042494.1 | NP_001035959.1 | 16 | 2381 | ||
NM_001042495.1 | NP_001035960.1 | 16 | 2056 | |||
NM_001042496.1 | NP_001035961.1 | 16 | 2447 | |||
NM_001042497.1 | NP_001035962.1 | 14 | 2150 | |||
NM_005135.2 | NP_005126.1 | 15 | 2020 | |||
NM_133647.1 | NP_598408.1 | 15 | 2195 | |||
XM_006720793.3 | XP_006720856.1 | 14 | 2930 | |||
XM_011522269.2 | XP_011520571.1 | 15 | 3077 | |||
GenBank mRNA | AF105366.1 | - | 15 | 2195 | ||
AF108831.1 | - | 15 | 1929 | |||
AF116242.1 | - | 15 | 2031 | |||
AF477977.1 | - | 16 | 2388 | |||
AF531258.1 | - | 14 | 2150 | |||
AF531259.1 | - | 16 | 2388 | |||
AF531260.1 | - | 16 | 2026 | |||
AK128133.1 | - | 14 | 1703 | |||
AK292550.1 | - | 16 | 2134 | |||
AK315283.1 | - | 15 | 1992 | |||
AL117500.1 | - | 12 | 1623 | |||
BC051744.1 | - | 16 | 2384 | 4829214 | ||
BC070107.1 | - | 16 | 2051 | 4826538 | ||
BC126241.1 | - | 15 | 1928 | 8991957 | ||
BC126243.1 | - | 15 | 1928 | 8991959 | ||
BX648195.1 | - | 15 | 3137 | |||
DQ138323.1 | - | 16 | 2446 |