Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169109.1 | NP_001162580.1 | 2 | 952 | ||
NM_001169110.1 | NP_001162581.1 | 2 | 903 | |||
NM_001169111.1 | NP_001162582.1 | 2 | 921 | |||
NM_005138.2 | NP_005129.2 | 2 | 892 | |||
GenBank mRNA | AL021683.1 | - | 2 | 830 | ||
BC102024.1 | - | 2 | 820 | 40030584 | ||
BC102025.1 | - | 2 | 820 | 40030589 | ||
CR456569.1 | - | 1 | 750 |