Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000479.3 | NP_000470.2 | 1 | 334 | ||
GenBank mRNA | BC049194.1 | - | 1 | 137 | 5168299 |