Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001181.4 | NP_001172.1 | 3 | 497 | ||
NM_001201352.1 | NP_001188281.1 | 3 | 497 | |||
NM_080912.3 | NP_550434.1 | 3 | 585 | |||
NM_080913.3 | NP_550435.1 | 3 | 440 | |||
NM_080914.2 | NP_550436.1 | 2 | 243 | |||
XM_005256648.2 | XP_005256705.1 | 4 | 550 | |||
XM_006721526.1 | XP_006721589.1 | 4 | 997 | |||
XM_011523862.2 | XP_011522164.2 | 4 | 567 | |||
XM_011523863.2 | XP_011522165.1 | 3 | 436 | |||
XM_011523864.2 | XP_011522166.1 | 3 | 471 | |||
XM_011523865.1 | XP_011522167.1 | 4 | 997 | |||
XM_011523866.1 | XP_011522168.1 | 4 | 547 | |||
XM_017024651.1 | XP_016880140.1 | 3 | 436 | |||
XM_017024652.1 | XP_016880141.1 | 4 | 966 | |||
XM_017024653.1 | XP_016880142.1 | 4 | 1054 | |||
GenBank mRNA | AF529374.1 | - | 2 | 230 | ||
AK303787.1 | - | 4 | 556 | |||
AK310123.1 | - | 3 | 542 | |||
BC017251.2 | - | 3 | 388 | 5087419 | ||
BX483468.1 | - | 3 | 453 | |||
DC315727.1 | - | 4 | 552 | |||
M11025.1 | - | 3 | 350 | |||
U97197.1 | - | 2 | 173 | |||
X55283.1 | - | 3 | 363 | |||
X55284.1 | - | 3 | 427 |