Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000754.3 | NP_000745.1 | 6 | 1281 | ||
NM_001135161.1 | NP_001128633.1 | 6 | 1239 | |||
NM_001135162.1 | NP_001128634.1 | 6 | 1256 | |||
NM_007310.2 | NP_009294.1 | 4 | 1012 | |||
XM_011529886.1 | XP_011528188.1 | 6 | 1186 | |||
XM_017028594.1 | XP_016884083.1 | 6 | 1505 | |||
XM_017028595.1 | XP_016884084.1 | 6 | 1449 | |||
GenBank mRNA | AK130031.1 | - | 7 | 1062 | ||
AK290440.1 | - | 6 | 1220 | |||
BC011935.2 | - | 5 | 1125 | 3503220 | ||
BC100018.1 | - | 6 | 1271 | 6163659 | ||
M65213.1 | - | 1 | 282 |