Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194392
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
33
3035
NM_001081752.2
NP_001075221.1
30
2894
NM_001081753.2
NP_001075222.1
31
2939
NM_001081754.2
NP_001075223.1
32
2996
NM_001081755.2
NP_001075224.1
32
2978
NM_001278912.1
NP_001265841.1
32
2981
NM_001278913.1
NP_001265842.1
29
2792
NM_001278914.1
NP_001265843.1
31
2963
NM_001278915.1
NP_001265844.1
33
3053
NM_001278916.1
NP_001265845.1
29
2837
NM_001278917.1
NP_001265846.1
32
3005
NM_001278918.1
NP_001265847.1
28
2714
NM_001278939.1
NP_001265868.1
34
3221
XM_005250187.1
XP_005250244.1
32
2676
XM_005250188.1
XP_005250245.1
32
2668
XM_011515868.1
XP_011514170.1
33
2736
XM_011515869.1
XP_011514171.1
32
2665
XM_011515870.1
XP_011514172.1
32
3031
XM_011515871.1
XP_011514173.1
32
2694
XM_011515872.1
XP_011514174.1
32
2682
XM_011515873.1
XP_011514175.1
32
2679
XM_011515874.1
XP_011514176.1
31
3001
XM_011515875.1
XP_011514177.1
31
2657
XM_011515876.1
XP_011514178.1
31
2637
XM_011515877.1
XP_011514179.1
31
2625
XM_017011813.1
XP_016867302.1
31
2579
XM_017011814.1
XP_016867303.1
31
2637
GenBank mRNA
AB208942.1
-
32
2631
AK075554.1
-
29
2475
AK092232.1
-
28
2329
AK094138.1
-
33
2468
AK095990.1
-
31
2559
AK122731.1
-
25
2187
AK125511.1
-
31
2621
AK130894.1
-
18
1530
AK225659.1
-
32
2592
BC035570.1
-
33
2640
BC050379.1
-
26
1909
BC065566.1
-
29
2468
6153564
BX537939.1
-
32
2616
BX538199.1
-
30
2526
M26867.1
-
1
381
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline