Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204502.1 | NP_001191431.1 | 9 | 1023 | ||
NM_001204503.1 | NP_001191432.1 | 8 | 992 | |||
NM_001278637.1 | NP_001265566.1 | 9 | 1000 | |||
NM_001278638.1 | NP_001265567.1 | 9 | 989 | |||
NM_001459.3 | NP_001450.2 | 9 | 972 | |||
XM_005258680.4 | XP_005258737.3 | 8 | 908 | |||
XM_005258681.4 | XP_005258738.3 | 8 | 959 | |||
XM_005258682.4 | XP_005258739.3 | 7 | 875 | |||
XM_005258683.4 | XP_005258740.3 | 7 | 823 | |||
XM_011526675.2 | XP_011524977.1 | 8 | 829 | |||
XM_011526676.2 | XP_011524978.1 | 8 | 794 | |||
XM_011526678.2 | XP_011524980.1 | 8 | 775 | |||
XM_011526682.2 | XP_011524984.1 | 8 | 979 | |||
XM_017026533.1 | XP_016882022.1 | 7 | 719 | |||
XM_017026534.1 | XP_016882023.1 | 7 | 692 | |||
XM_017026535.1 | XP_016882024.1 | 9 | 941 | |||
GenBank mRNA | BC006331.2 | - | 9 | 962 | ||
BC011914.2 | - | 9 | 962 | |||
BC028001.1 | - | 9 | 973 | 5198812 | ||
U03858.1 | - | 8 | 996 |