Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000805.4 | NP_000796.1 | 3 | 363 | ||
GenBank mRNA | BC069724.1 | - | 3 | 328 | 7262518 | |
BC069762.1 | - | 3 | 328 | 7262530 | ||
BM768483.1 | - | 3 | 362 | |||
BM768762.1 | - | 3 | 364 | |||
V00511.1 | - | 2 | 304 |