Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122951.2 | NP_001116423.1 | 1 | 1217 | ||
NM_002036.3 | NP_002027.2 | 2 | 1983 | |||
GenBank mRNA | AK291593.1 | - | 2 | 1983 | ||
BC017817.1 | - | 2 | 1188 | 4696425 | ||
BX537430.1 | - | 3 | 1880 | |||
U01839.1 | - | 1 | 1217 |